Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032556.5 | NP_115945.4 | 4 | 896 | |
NM_173161.2 | NP_775184.1 | 5 | 550 | ||
GenBank mRNA | AF334755.1 | - | 4 | 896 | |
AY026753.1 | - | 2 | 541 | ||
AY029413.1 | - | 5 | 550 | ||
AY358846.1 | - | 5 | 538 | ||
BC103966.1 | - | 5 | 541 | 40014540 | |
BC103967.1 | - | 5 | 541 | 40014541 | |
BC103968.1 | - | 5 | 541 | 40014542 |