Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr. 14: 100376410 - 100530303 on Build GRCh38
UniGene ID
Hs.497600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161476.1 NP_001154948.1 5 1318
NM_024515.4 NP_078791.3 5 1449
XM_005268056.4 XP_005268113.1 5 1325
XM_005268057.3 XP_005268114.1 4 475
XM_005268058.4 XP_005268115.1 4 489
XM_011537150.2 XP_011535452.1 5 1572
XM_011537151.2 XP_011535453.2 4 928
XM_017021639.1 XP_016877128.1 4 477
GenBank mRNA AK291960.1 - 5 1281
AY295082.1 - 5 1332
BC003641.2 - 4 555 3529568
BC007953.2 - 3 1102
CT003969.1 - 4 481

View Full Product Details