Gene Transcripts

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
Chromosome Location
Chr. 1: 2050470 - 2185395 on Build GRCh38
UniGene ID
Hs.496255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002744.4 NP_002735.3 2 295
XM_011541773.1 XP_011540075.1 2 291
XM_011541775.2 XP_011540077.1 2 291
XM_017001789.1 XP_016857278.1 2 291
XM_017001790.1 XP_016857279.1 2 291
XM_017001803.1 XP_016857292.1 2 291
GenBank mRNA AK290995.1 - 2 134
BC008058.2 - 2 242 2987996
BC014270.2 - 2 295 3835020
BT007082.1 - 2 134
L14283.1 - 2 134
Z15108.1 - 2 116

View Full Product Details