Gene Transcripts

Gene Symbol
Il6st
Entrez Gene ID
16195
Gene Name
interleukin 6 signal transducer
Gene Aliases
5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
Chromosome Location
Chr. 13: 112464070 - 112506860 on Build GRCm38
UniGene ID
Mm.4364
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010560.3 NP_034690.3 6 816
GenBank mRNA BC020454.1 - 6 770
BC058679.1 - 6 777 6834623
M83336.1 - 5 621
X62646.1 - 5 621

View Full Product Details