Gene Transcripts
- Gene Symbol
- ZNF232
- Entrez Gene ID
- 7775
- Gene Name
- zinc finger protein 232
- Gene Aliases
- ZSCAN11
- Chromosome Location
- Chr. 17: 5105734 - 5123116 on Build GRCh38
- UniGene ID
- Hs.279914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC002852.1 |
- |
|
1703 |
|
- Gene Symbol
- CFAP36
- Entrez Gene ID
- 112942
- Gene Name
- cilia and flagella associated protein 36
- Gene Aliases
- BARTL1, CCDC104
- Chromosome Location
- Chr. 2: 55519595 - 55545080 on Build GRCh38
- UniGene ID
- Hs.264208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC001797.1 |
- |
|
1328 |
|
View Full Product Details