Gene Transcripts
- Gene Symbol
- WDR25
- Entrez Gene ID
- 79446
- Gene Name
- WD repeat domain 25
- Gene Aliases
- C14orf67
- Chromosome Location
- Chr. 14: 100376410 - 100530303 on Build GRCh38
- UniGene ID
- Hs.497600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC003641.2 |
- |
1 |
116 |
|
View Full Product Details