Gene Transcripts

Gene Symbol
Btbd9
Entrez Gene ID
294318
Gene Name
BTB (POZ) domain containing 9
Gene Aliases
Chromosome Location
Chr. 20: 8899286 - 9253931 on Build Rnor_6.0
UniGene ID
Rn.48897
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013073.1 NP_001013091.1 9 1586
XM_006256185.2 XP_006256247.1 9 1612
GenBank mRNA BC087068.1 - 9 1586

View Full Product Details