Gene Transcripts
- Gene Symbol
- FAM87A
- Entrez Gene ID
- 157693
- Gene Name
- family with sequence similarity 87 member A
- Gene Aliases
- Chromosome Location
- Chr. 8: 375931 - 383174 on Build GRCh38
- UniGene ID
- Hs.591390
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103537.1 |
|
4 |
3824 |
|
GenBank mRNA |
BC037297.2 |
- |
4 |
3823 |
|
- Gene Symbol
- FAM87B
- Entrez Gene ID
- 400728
- Gene Name
- family with sequence similarity 87 member B
- Gene Aliases
- Chromosome Location
- Chr. 1: 817371 - 819834 on Build GRCh38
- UniGene ID
- Hs.568078
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103536.1 |
|
2 |
1758 |
|
GenBank mRNA |
AK097327.1 |
- |
2 |
1758 |
|
View Full Product Details