Gene Transcripts

Gene Symbol
CFC1
Entrez Gene ID
55997
Gene Name
cripto, FRL-1, cryptic family 1
Gene Aliases
CFC1B, CRYPTIC, DTGA2, HTX2
Chromosome Location
Chr. 2: 130592165 - 130599575 on Build GRCh38
UniGene ID
Hs.567542
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270420.1 NP_001257349.1 2, 3 370
NM_001270421.1 NP_001257350.1 2, 3 370
NM_032545.3 NP_115934.1 2, 3 370
XM_011511486.2 XP_011509788.1 2, 3 365
GenBank mRNA AF312769.1 - 2, 3 128
AK290094.1 - 2, 3 346
AK315326.1 - 2, 3 370
BC069508.1 - 2, 3 152 7262189
BC074825.2 - 2, 3 131 30915323
BC074826.2 - 2, 3 131 30915540
BC110080.1 - 2, 3 243 40030641
BC146897.1 - 2, 3 247 9056825
BM509218.1 - 2, 3 304
CA777249.1 - 2, 3 343
CA941621.1 - 2, 3 302

Gene Symbol
CFC1B
Entrez Gene ID
653275
Gene Name
cripto, FRL-1, cryptic family 1B
Gene Aliases
Chromosome Location
Chr. 2: 130521094 - 130527992 on Build GRCh38
UniGene ID
Hs.503733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079530.1 NP_001072998.1 2, 3 473
XM_005263737.4 XP_005263794.1 2, 3 355
GenBank mRNA BQ777283.1 - 2, 3 315
BU784123.1 - 2, 3 328

View Full Product Details