Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031326.1 | NP_112616.1 | 4, 5 | 547 | |
GenBank mRNA | AB014089.1 | - | 4, 5 | 547 | |
AF246196.1 | - | 3, 4 | 324 | ||
AF377866.1 | - | 4, 5 | 554 | ||
AJ312746.1 | - | 4, 5 | 547 | ||
BC062022.1 | - | 4, 5 | 549 | 5623491 | |
FQ219683.1 | - | 1, 2 | 104 |