Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027113.2 | NP_081389.1 | 1 | 649 | |
GenBank mRNA | AK009247.1 | - | 1 | 564 | |
AK149554.1 | - | 620 | |||
AK165100.1 | - | 513 | |||
BC064045.1 | - | 1 | 649 | 1245342 | |
EF011616.1 | - | 1 | 647 |