gRNA Design Details

Target DNA Sequence
TTCCGATGTAGCCACTGCCC
PAM Sequence
AGG
Target locus
Chr.2: 69642932 - 69642954 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.76/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
Chromosome Location
Chr.2: 69457995 - 69643845 on GRCh38
UniGene ID
Hs.468878
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_014911.3 NP_055726.3 2