Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001276468.1 | NP_001263397.1 | 3 |
NM_001276469.1 | NP_001263398.1 | 4 | |
NM_001478.4 | NP_001469.1 | 4 | |
XM_005268773.4 | XP_005268830.1 | 4 | |
XM_011538147.2 | XP_011536449.1 | 4 | |
XM_011538148.2 | XP_011536450.1 | 4 | |
XM_017019140.1 | XP_016874629.1 | 4 | |
XM_017019141.1 | XP_016874630.1 | 4 | |
XM_017019142.1 | XP_016874631.1 | 4 |