gRNA Design Details
- Target DNA Sequence
- TTTGAACGGCAGAGCCAGAA
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 16360328 - 16360350 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.44/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MGST1
- Entrez Gene ID
-
4257
- Gene Name
- microsomal glutathione S-transferase 1
- Gene Aliases
-
GST12, MGST, MGST-I
- Chromosome Location
-
Chr.12: 16347142 - 16377189 on GRCh38
- UniGene ID
- Hs.389700
- Species
-
Human