gRNA Design Details

Target DNA Sequence
CTTCCGAGCGCGTTGAGCTT
PAM Sequence
TGG
Target locus
Chr.11: 62591546 - 62591568 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 92.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TUT1
Entrez Gene ID
64852
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
PAPD2, RBM21, STARPAP, TUTase, URLC6
Chromosome Location
Chr.11: 62575045 - 62591637 on GRCh38
UniGene ID
Hs.728983
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_022830.2 NP_073741.2 1