gRNA Design Details
- Target DNA Sequence
- TCGGTACAGGAGAGTCGCTA
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 62591482 - 62591504 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TUT1
- Entrez Gene ID
-
64852
- Gene Name
- terminal uridylyl transferase 1, U6 snRNA-specific
- Gene Aliases
-
PAPD2, RBM21, STARPAP, TUTase, URLC6
- Chromosome Location
-
Chr.11: 62575045 - 62591637 on GRCh38
- UniGene ID
- Hs.728983
- Species
-
Human