gRNA Design Details
- Target DNA Sequence
- CCAGCTGTTCAAGAATGGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 58353072 - 58353094 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- A1BG
- Entrez Gene ID
-
1
- Gene Name
- alpha-1-B glycoprotein
- Gene Aliases
-
A1B, ABG, GAB, HYST2477
- Chromosome Location
-
Chr.19: 58346806 - 58353499 on GRCh38
- UniGene ID
- Hs.529161
- Species
-
Human