gRNA Design Details
- Target DNA Sequence
- GCCCGTATTGGCCATGGCAT
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 50214732 - 50214754 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SLC38A3
- Entrez Gene ID
-
10991
- Gene Name
- solute carrier family 38 member 3
- Gene Aliases
-
G17, NAT1, SN1, SNAT3
- Chromosome Location
-
Chr.3: 50205246 - 50220979 on GRCh38
- UniGene ID
- Hs.76460
- Species
-
Human