gRNA Design Details
- Target DNA Sequence
- CACATGGAGGCTGCTCATTT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 61916092 - 61916114 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Akap10
- Entrez Gene ID
-
56697
- Gene Name
- A kinase (PRKA) anchor protein 10
- Gene Aliases
-
1500031L16Rik, B130049N18Rik, D-AKAP-2, D-AKAP2, PRKA10
- Chromosome Location
-
Chr.11: 61871307 - 61930307 on GRCm38
- UniGene ID
- Mm.274404
- Species
-
Mouse