gRNA Design Details
- Target DNA Sequence
- GACCTACTCGAACGTGGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 58128133 - 58128155 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hnrnpa0
- Entrez Gene ID
-
77134
- Gene Name
- heterogeneous nuclear ribonucleoprotein A0
- Gene Aliases
-
1110055B05Rik, 3010025E17Rik, Hnrpa0, hnRNP A0
- Chromosome Location
-
Chr.13: 58125879 - 58128556 on GRCm38
- UniGene ID
- Mm.490274
- Species
-
Mouse