gRNA Design Details
- Target DNA Sequence
- CAAGAAAATTGTTGGCTAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 52750341 - 52750363 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTA2
- Entrez Gene ID
-
2939
- Gene Name
- glutathione S-transferase alpha 2
- Gene Aliases
-
GST2, GSTA2-2, GTA2, GTH2
- Chromosome Location
-
Chr.6: 52750087 - 52763563 on GRCh38
- UniGene ID
- Hs.94107
- Species
-
Human