gRNA Design Details
- Target DNA Sequence
- GTCATACCGCTTTTCCCTGG
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 60920844 - 60920866 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- A1bg
- Entrez Gene ID
-
117586
- Gene Name
- alpha-1-B glycoprotein
- Gene Aliases
-
C44
- Chromosome Location
-
Chr.15: 60917589 - 60921270 on GRCm38
- UniGene ID
- Mm.291499
- Species
-
Mouse