gRNA Design Details
- Target DNA Sequence
- AGCAACAACCACTTCCGAGG
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 129830744 - 129830766 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gar1
- Entrez Gene ID
-
68147
- Gene Name
- GAR1 ribonucleoprotein homolog (yeast)
- Gene Aliases
-
AA409823, AI326794, C430047J18Rik, Nola1
- Chromosome Location
-
Chr.3: 129824912 - 129831396 on GRCm38
- UniGene ID
- Mm.24873
- Species
-
Mouse