gRNA Design Details
- Target DNA Sequence
- CTATGGTACGCAGCTGGAAG
- PAM Sequence
- GGG
- Target locus
-
Chr.10: 75780859 - 75780881 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstt3
- Entrez Gene ID
-
103140
- Gene Name
- glutathione S-transferase, theta 3
- Gene Aliases
-
AI118089
- Chromosome Location
-
Chr.10: 75774122 - 75781483 on GRCm38
- UniGene ID
- Mm.5731
- Species
-
Mouse