gRNA Design Details
- Target DNA Sequence
- GTTTTCATATCGGAAGTGGG
- PAM Sequence
- TGG
- Application
- Gene Knockout
Score:
88.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Satl1
- Entrez Gene ID
-
73809
- Gene Name
- spermidine/spermine N1-acetyl transferase-like 1
- Gene Aliases
-
4930404K22Rik
- Chromosome Location
-
Chr.X: 112383207 - 112406779 on GRCm38
- UniGene ID
- Mm.50508
- Species
-
Mouse