gRNA Design Details

Target DNA Sequence
GTTGGCACATCGAGAATATC
PAM Sequence
AGG
Application
Gene Knockout
Score: 72.02/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
OGT
Entrez Gene ID
8473
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
Gene Aliases
HINCUT-1, HRNT1, O-GLCNAC
Chromosome Location
Chr.X: 71533062 - 71575897 on GRCh38
UniGene ID
Hs.405410
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_181672.2 NP_858058.1 2
NM_181673.2 NP_858059.1 2
XM_017029907.1 XP_016885396.1 1
XM_017029908.1 XP_016885397.1 2