gRNA Design Details
- Target DNA Sequence
- AGACAAGAGCCTGACAATAC
- PAM Sequence
- TGG
- Application
- Gene Knockout
Score:
82.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ogt
- Entrez Gene ID
-
108155
- Gene Name
- O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)
- Gene Aliases
-
1110038P24Rik, 4831420N21Rik, AI115525, Ogtl
- Chromosome Location
-
Chr.X: 101640012 - 101684351 on GRCm38
- UniGene ID
- Mm.487332
- Species
-
Mouse