gRNA Design Details
- Target DNA Sequence
- CTACTCATACCCCACCAAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 96698628 - 96698650 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Slc38a2
- Entrez Gene ID
-
67760
- Gene Name
- solute carrier family 38, member 2
- Gene Aliases
-
5033402L14Rik, AI316867, ATA2, SAT2, mKIAA1382
- Chromosome Location
-
Chr.15: 96687386 - 96699724 on GRCm38
- UniGene ID
- Mm.46754
- Species
-
Mouse