gRNA Design Details
- Target DNA Sequence
- AAACATAGCGGCGGATAAGG
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 123116366 - 123116388 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
73.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Oxct2b
- Entrez Gene ID
-
353371
- Gene Name
- 3-oxoacid CoA transferase 2B
- Gene Aliases
-
Scot-t2
- Chromosome Location
-
Chr.4: 123116248 - 123118000 on GRCm38
- UniGene ID
- Mm.189660
- Species
-
Mouse