gRNA Design Details
- Target DNA Sequence
- TGACTTCGATGCCATGGATC
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 70035158 - 70035180 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gcnt3
- Entrez Gene ID
-
72077
- Gene Name
- glucosaminyl (N-acetyl) transferase 3, mucin type
- Gene Aliases
-
2010013H22Rik, 2210021I22Rik, 2210401J11Rik
- Chromosome Location
-
Chr.9: 70031496 - 70039887 on GRCm38
- UniGene ID
- Mm.195555
- Species
-
Mouse