gRNA Design Details
- Target DNA Sequence
- AGACATTCCTTGCTTAAAGG
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 47876399 - 47876421 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gsto2
- Entrez Gene ID
-
68214
- Gene Name
- glutathione S-transferase omega 2
- Gene Aliases
-
1700020F09Rik, 4930425C18Rik, GSTO 2-2
- Chromosome Location
-
Chr.19: 47865545 - 47886308 on GRCm38
- UniGene ID
- Mm.63791
- Species
-
Mouse