gRNA Design Details
- Target DNA Sequence
- GGCTGGCTTCCTGATGTTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 25240075 - 25240097 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
80.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gnptg
- Entrez Gene ID
-
214505
- Gene Name
- N-acetylglucosamine-1-phosphotransferase, gamma subunit
- Gene Aliases
-
6430527N14Rik, A830081F19, AU067667, AU067744, Mdcp1, Tce7
- Chromosome Location
-
Chr.17: 25234317 - 25240170 on GRCm38
- UniGene ID
- Mm.38607
- Species
-
Mouse