gRNA Design Details
- Target DNA Sequence
- CCGTAGTATCGGTGGGACTC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 120294449 - 120294471 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- B4galnt3
- Entrez Gene ID
-
330406
- Gene Name
- beta-1,4-N-acetyl-galactosaminyl transferase 3
- Gene Aliases
-
AB114826, B4GalNAcT3, C330047A21
- Chromosome Location
-
Chr.6: 120200753 - 120295005 on GRCm38
- UniGene ID
- Mm.321840
- Species
-
Mouse