gRNA Design Details
- Target DNA Sequence
- GTGTGCTCATCCGGAAACAG
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 122575453 - 122575475 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.81/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aprt
- Entrez Gene ID
-
11821
- Gene Name
- adenine phosphoribosyl transferase
- Gene Aliases
-
C85684
- Chromosome Location
-
Chr.8: 122574637 - 122576907 on GRCm38
- UniGene ID
- Mm.1786
- Species
-
Mouse