gRNA Design Details
- Target DNA Sequence
- CGAGTATGTGGCCGACTTCT
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 120018847 - 120018869 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aatk
- Entrez Gene ID
-
11302
- Gene Name
- apoptosis-associated tyrosine kinase
- Gene Aliases
-
AATYK, aatyk1, mKIAA0641
- Chromosome Location
-
Chr.11: 120007311 - 120047182 on GRCm38
- UniGene ID
- Mm.6826
- Species
-
Mouse