gRNA Design Details
- Target DNA Sequence
- TGACAGTATGAAGCCCCTGG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 17672576 - 17672598 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sugct
- Entrez Gene ID
-
192136
- Gene Name
- succinyl-CoA glutarate-CoA transferase
- Gene Aliases
-
5033411D12Rik, D17907
- Chromosome Location
-
Chr.13: 16857472 - 17694793 on GRCm38
- UniGene ID
- Mm.364180
- Species
-
Mouse