gRNA Design Details
- Target DNA Sequence
- GGGTGACATATTTGCCGCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 4036705 - 4036727 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstp1
- Entrez Gene ID
-
14870
- Gene Name
- glutathione S-transferase, pi 1
- Gene Aliases
-
GstpiB
- Chromosome Location
-
Chr.19: 4035411 - 4037912 on GRCm38
- UniGene ID
- Mm.299292
- Species
-
Mouse