gRNA Design Details
- Target DNA Sequence
- ACAGACTCGAGCCAGCTGAT
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 108043339 - 108043361 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.41/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstm4
- Entrez Gene ID
-
14865
- Gene Name
- glutathione S-transferase, mu 4
- Gene Aliases
-
1110004G14Rik, GSTM7-7, Gstb-4, Gstb4
- Chromosome Location
-
Chr.3: 108040408 - 108045080 on GRCm38
- UniGene ID
- Mm.31203
- Species
-
Mouse