gRNA Design Details
- Target DNA Sequence
- GTACCAGCCATGATGGATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 75797061 - 75797083 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstt1
- Entrez Gene ID
-
14871
- Gene Name
- glutathione S-transferase, theta 1
- Gene Aliases
-
AI255817, Gstt1-1
- Chromosome Location
-
Chr.10: 75783813 - 75798584 on GRCm38
- UniGene ID
- Mm.2746
- Species
-
Mouse