gRNA Design Details
- Target DNA Sequence
- GGGGACATAAAAGTTATTGG
- PAM Sequence
- TGG
- Application
- Gene Knockout
Score:
82.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hprt
- Entrez Gene ID
-
15452
- Gene Name
- hypoxanthine guanine phosphoribosyl transferase
- Gene Aliases
-
C81579, HPGRT, Hprt1
- Chromosome Location
-
Chr.X: 52988078 - 53021660 on GRCm38
- UniGene ID
- Mm.299381
- Species
-
Mouse