gRNA Design Details
- Target DNA Sequence
- AATTCGAACTCTCCATGAAC
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 87157921 - 87157943 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstz1
- Entrez Gene ID
-
14874
- Gene Name
- glutathione transferase zeta 1 (maleylacetoacetate isomerase)
- Gene Aliases
-
MAAI
- Chromosome Location
-
Chr.12: 87147165 - 87164723 on GRCm38
- UniGene ID
- Mm.29652
- Species
-
Mouse