gRNA Design Details
- Target DNA Sequence
- CTGTTCCCGCTCTAGCCTTT
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 55721640 - 55721662 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
82.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rabggta
- Entrez Gene ID
-
56187
- Gene Name
- Rab geranylgeranyl transferase, a subunit
- Gene Aliases
-
gm
- Chromosome Location
-
Chr.14: 55715877 - 55722368 on GRCm38
- UniGene ID
- Mm.87216
- Species
-
Mouse