gRNA Design Details
- Target DNA Sequence
- GAGGATCCGTGCAGACATTG
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 108016387 - 108016409 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
71.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gstm1
- Entrez Gene ID
-
14862
- Gene Name
- glutathione S-transferase, mu 1
- Gene Aliases
-
Gstb-1, Gstb1
- Chromosome Location
-
Chr.3: 108012250 - 108017973 on GRCm38
- UniGene ID
- Mm.37199
- Species
-
Mouse