gRNA Design Details
- Target DNA Sequence
- TCATACGTTCTTCACGACGG
- PAM Sequence
- GGG
- Target locus
-
Chr.13: 40918041 - 40918063 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gcnt2
- Entrez Gene ID
-
14538
- Gene Name
- glucosaminyl (N-acetyl) transferase 2, I-branching enzyme
- Gene Aliases
-
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC
- Chromosome Location
-
Chr.13: 40859768 - 40960892 on GRCm38
- UniGene ID
- Mm.314757
- Species
-
Mouse