gRNA Design Details
- Target DNA Sequence
- AAACTCGGGATGACCTGGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 21257350 - 21257372 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gsta3
- Entrez Gene ID
-
14859
- Gene Name
- glutathione S-transferase, alpha 3
- Gene Aliases
-
Gst2-3
- Chromosome Location
-
Chr.1: 21240585 - 21265575 on GRCm38
- UniGene ID
- Mm.394593
- Species
-
Mouse