gRNA Design Details
- Target DNA Sequence
- GTCTAGCAGATCGAGTCTCG
- PAM Sequence
- GGG
- Target locus
-
Chr.10: 127166141 - 127166163 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- B4galnt1
- Entrez Gene ID
-
14421
- Gene Name
- beta-1,4-N-acetyl-galactosaminyl transferase 1
- Gene Aliases
-
4933429D13Rik, Gal-NAc-T, GalNAc-T, GalNAcT, Galgt1, Ggm-2, Ggm2
- Chromosome Location
-
Chr.10: 127165156 - 127172340 on GRCm38
- UniGene ID
- Mm.386762
- Species
-
Mouse