gRNA Design Details

Target DNA Sequence
GCAGCAAGACTAGCGCATAG
PAM Sequence
AGG
Target locus
Chr.10: 127166025 - 127166047 on GRCm38
Strand
Reverse
Application
Gene Knockout
Score: 88.52/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
B4galnt1
Entrez Gene ID
14421
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 1
Gene Aliases
4933429D13Rik, Gal-NAc-T, GalNAc-T, GalNAcT, Galgt1, Ggm-2, Ggm2
Chromosome Location
Chr.10: 127165156 - 127172340 on GRCm38
UniGene ID
Mm.386762
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001244617.1 NP_001231546.1 2
NM_001244618.1 NP_001231547.1 1
NM_008080.5 NP_032106.1 2
NM_027739.2 NP_082015.2 1
NR_045166.1 2