gRNA Design Details
- Target DNA Sequence
- GCAACCAGGCAGTGGTAGAA
- PAM Sequence
- GGG
- Application
- Gene Knockout
Score:
78.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sat1
- Entrez Gene ID
-
20229
- Gene Name
- spermidine/spermine N1-acetyl transferase 1
- Gene Aliases
-
AA617398, SSAT, Sat
- Chromosome Location
-
Chr.X: 155213126 - 155216449 on GRCm38
- UniGene ID
- Mm.2734
- Species
-
Mouse