gRNA Design Details
- Target DNA Sequence
- GCCTTCTTGTACGGGTCATC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 47858025 - 47858047 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gsto1
- Entrez Gene ID
-
14873
- Gene Name
- glutathione S-transferase omega 1
- Gene Aliases
-
AA407097, AI194287, AU018802, GSTO 1-1, GSTX, SPG-R, p28
- Chromosome Location
-
Chr.19: 47854989 - 47864790 on GRCm38
- UniGene ID
- Mm.378931
- Species
-
Mouse