gRNA Design Details
- Target DNA Sequence
- CATCCCGTCAATTTCAACCA
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 52985534 - 52985556 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTA4
- Entrez Gene ID
-
2941
- Gene Name
- glutathione S-transferase alpha 4
- Gene Aliases
-
GSTA4-4, GTA4
- Chromosome Location
-
Chr.6: 52977948 - 52995380 on GRCh38
- UniGene ID
- Hs.485557
- Species
-
Human